Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sch 563705 | 473728-58-4 | 425.48 g/mol | C23H27N3O5 | 5 MG
SDP

Catalog No. 5000215150
Encompass_Preferred

(Rac)-SCH 563705 is the racemic form of SCH 563705, a small-molecule antagonist of chemokine receptors CXCR1 and CXCR2 used for research applications. Supplied as a low-mass solid for laboratory use, it has documented nanomolar potency and defined chemical identifiers for unambiguous reference.

  • Racemic small-molecule antagonist of CXCR1 and CXCR2.
  • Demonstrated nanomolar potency in receptor assays.
  • Molecular formula C23H27N3O5.
  • Molecular weight 425.48 g/mol.
  • Provided as a 5 mg solid suitable for in vitro and in vivo research.

Catalog No. 50-002-15150 Supplier Medchemexpress LLC Supplier No. HY10011B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-SCH 563705 is the racemic form of SCH 563705, a small-molecule antagonist of chemokine receptors CXCR1 and CXCR2 used for research applications. Supplied as a low-mass solid for laboratory use, it has documented nanomolar potency and defined chemical identifiers for unambiguous reference.

  • Racemic small-molecule antagonist of CXCR1 and CXCR2.
  • Demonstrated nanomolar potency in receptor assays.
  • Molecular formula C23H27N3O5.
  • Molecular weight 425.48 g/mol.
  • Provided as a 5 mg solid suitable for in vitro and in vivo research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.