Learn More
Cayman Chemical SCH 79797 (hydrochloride), Purity: ≥98%, CAS: 1216720-69-2, 5 mg
SDP
Catalog No. 502971464
Encompass_Preferred
A non-peptide antagonist of PAR1; blocks binding of haTRAP (IC50 = 70 nM) as well as platelet aggregation induced by either haTRAP or α-thrombin (IC50 = 0.3 and 3 µM, respectively); has no activity against PAR2, PAR4, or other receptors involved in platelet activation
Catalog No. 50-297-1464 Supplier Cayman Chemical Supplier No. 146335 MG
A non-peptide antagonist of PAR1; blocks binding of haTRAP (IC50 = 70 nM) as well as platelet aggregation induced by either haTRAP or α-thrombin (IC50 = 0.3 and 3 µM, respectively); has no activity against PAR2, PAR4, or other receptors involved in platelet activation