Learn More
NEOGEN Schaedler Agar, 500 g
SDP
Catalog No. 502015597
Encompass_Preferred
Schaedler Agar is used for the cultivation of anaerobic microorganisms in a laboratory setting. Schaedler Agar is not intended for use in the diagnosis of disease or other conditions in humans. Survival of anaerobic bacteria is dependent on their sensitivity to oxygen, nutritional requirements, appropriate collection, culture medium, and incubation time and temperature. Schaedler Agar is suitable for standard procedures used in cultivating anaerobic bacteria. Schaedler Agar is prepared according to the formulation described by Schaedler, Dubos, and Costello, and modified by Mata, Carrillo, and Villatoro. Modifications include reduced dextrose to avoid interference with hemolytic reactions, reduced yeast extract to avoid darkening of the medium, and adjusted sodium chloride and nitrogen concentrations.For more information visit https://www.neogen.com/solutions/Microbiology/schaedler-agar
Catalog No. 50-201-5597 Supplier NEOGEN Supplier No. 700003412
Schaedler Agar is used for the cultivation of anaerobic microorganisms in a laboratory setting. Schaedler Agar is not intended for use in the diagnosis of disease or other conditions in humans. Survival of anaerobic bacteria is dependent on their sensitivity to oxygen, nutritional requirements, appropriate collection, culture medium, and incubation time and temperature. Schaedler Agar is suitable for standard procedures used in cultivating anaerobic bacteria. Schaedler Agar is prepared according to the formulation described by Schaedler, Dubos, and Costello, and modified by Mata, Carrillo, and Villatoro. Modifications include reduced dextrose to avoid interference with hemolytic reactions, reduced yeast extract to avoid darkening of the medium, and adjusted sodium chloride and nitrogen concentrations.For more information visit https://www.neogen.com/solutions/Microbiology/schaedler-agar