Learn More
Medchemexpress LLC HY-103169 5mg Medchemexpress, SCH442416 CAS:316173-57-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1031695MG
Encompass_Preferred
Medchemexpress, HY-103169 5mg SCH442416 CAS:316173-57-6 SCH442416 is a potent, selective and brain-penetrant antagonist of adenosine A2A receptor (A2AR), with Kis of 0.048 and 0.5 nM for human and rat A2AR respectively. SCH442416 displays more than 23000-fold selectivity over A1R, A2BR, and A3R (Ki=1111, 10000, and 10000 nM, respectively). SCH442416 can be used for imaging of adenosine A2A receptors in rat and primate brain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0658