Learn More
Selleck Chemical LLC SCH58261 5mg
SDP
Supplier:  Selleck Chemical LLC S81045MG
Encompass_Preferred
SCH 58261 is a potent and selective A2a adenosine receptor antagonist with Ki of 2.3 nM and 2 nM for rat A2a and bovine A2a, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3340