Learn More
Apexbio Technology LLC SCH772984, 10mM (in 1mL DMSO) Cas No: 942183-80-4 MFCD: MFCD11878503
SDP
Supplier:  Apexbio Technology LLC A38055.1
Encompass_Preferred
SCH772984, identified by an affinity-based mass spectroscopy high-throughput platform, is a novel, potent and ATP-competitive inhibition of ERK1 and ERK2 with 50% inhibition concentration IC50 values of 4 nmol/L and 1 nmol/L respectively. Although it displays behaviors of both type I and type II kinase inhibitors, SCH772984 is highly selective against only seven kinases, including CLK2, FLT4, GSG2, MAP4K4, MAPK1, MINK1, PRKD1 and TTK, out of a wide range of 300 tested with more than 50% inhibition at a concentration of 1 μmol/L. Study results have shown that SCH772984 potently inhibits tumor cells with mutations in BRAF, NRAS and KRAS at nanomolar concentrations. Other sizes are available. Please inqury us for quote.
Catalog No. NC1482140