Learn More
Selleck Chemical LLC SCH772984 5mg 942183-80-4
SDP
Supplier:  Selleck Chemical LLC S71015MG
Encompass_Preferred
SCH772984 is a novel, specific inhibitor of ERK1/2 with IC50 values of 4 nM and 1 nM in cell-free assay, respectively, And show robust efficacy in RAS- or BRAF-mutant cancer cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1185170