Learn More
Cayman Chemical SCH 772984, 942183-80-4, 5 mg
SDP
Supplier:  Cayman Chemical 191665
Encompass_Preferred
A potent, selective inhibitor of ERK1 and ERK2 (IC50s = 4 and 1 nM, respectively); has nanomolar cytotoxicity in tumor cells with mutations in BRAF, NRAS, or KRAS; effective in vivo, inducing regression of xenograft tumors in mice
Catalog No. NC1490468