Learn More
Medchemexpress LLC SCH772984 | 942183-80-4 | 99.8% | C33H33N9O2 | 200 MG
SDP
Catalog No. 5000358341
Encompass_Preferred
SCH772984 is a highly selective and ATP-competitive ERK inhibitor, with IC50s of 4 nM for ERK1 and 1 nM for ERK2. It exhibits antitumor activity in both MAPK inhibitor-naive and MAPK inhibitor-resistant cells that contain BRAF or RAS mutations.
  • Highly selective and ATP-competitive ERK inhibitor
  • Exhibits antitumor activity in MAPK inhibitor-naive and resistant cells
  • Induces G1 arrest and apoptosis in sensitive melanoma cells
  • Inhibits ERK and RSK phosphorylation
  • Shows EC50 values less than 500 nM in BRAF-mutant and RAS-mutant tumor lines
  • Leads to significant tumor regression in animal models
  • Well tolerated in vivo
Catalog No. 50-003-58341 Supplier Medchemexpress LLC Supplier No. HY50846200MG
SCH772984 is a highly selective and ATP-competitive ERK inhibitor, with IC50s of 4 nM for ERK1 and 1 nM for ERK2. It exhibits antitumor activity in both MAPK inhibitor-naive and MAPK inhibitor-resistant cells that contain BRAF or RAS mutations.
  • Highly selective and ATP-competitive ERK inhibitor
  • Exhibits antitumor activity in MAPK inhibitor-naive and resistant cells
  • Induces G1 arrest and apoptosis in sensitive melanoma cells
  • Inhibits ERK and RSK phosphorylation
  • Shows EC50 values less than 500 nM in BRAF-mutant and RAS-mutant tumor lines
  • Leads to significant tumor regression in animal models
  • Well tolerated in vivo