Learn More
Apexbio Technology LLC SCH772984 TFA 942183-80-4 (free base) 5mg
SDP
Catalog No. 5000094310
Encompass_Preferred
SCH772984 TFA is a small-molecule inhibitor targeting extracellular signal-regulated kinase 1/2 (ERK1/2) It is designed to inhibit ERK catalytic activity thereby blocking phosphorylation of downstream substrates and regulating the RAS-RAF-MEK-ERK signaling pathway SCH772984 TFA exerts its biological activity primarily through direct interaction with ERK1/2 acting as an ATP-competitive inhibitor Based on these pharmacological properties SCH772984 TFA holds research potential in studies examining ERK signaling s role in oncogenic pathway activation tumor cell growth survival mechanisms and resistance phenomena in various cancer models
Catalog No. 50-000-94310 Supplier Apexbio Technology LLC Supplier No. B58675
SCH772984 TFA is a small-molecule inhibitor targeting extracellular signal-regulated kinase 1/2 (ERK1/2) It is designed to inhibit ERK catalytic activity thereby blocking phosphorylation of downstream substrates and regulating the RAS-RAF-MEK-ERK signaling pathway SCH772984 TFA exerts its biological activity primarily through direct interaction with ERK1/2 acting as an ATP-competitive inhibitor Based on these pharmacological properties SCH772984 TFA holds research potential in studies examining ERK signaling s role in oncogenic pathway activation tumor cell growth survival mechanisms and resistance phenomena in various cancer models