Learn More
TARGETMOL CHEMICALS INC SCH79797 DIHYDROCHLORIDE 5MG
SDP
Catalog No. 502581408
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. SCH79797 dihydrochloride is an effective and selective antagonist of protease activated receptor 1 (PAR1) with an IC50 of 70 nM and a Ki of 35 nM. SCH79797 dihydrochloride has antiproliferative and pro-apoptotic effects. purity: 98%
Catalog No. 50-258-1408 Supplier TARGETMOL CHEMICALS INC Supplier No. T128705MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. SCH79797 dihydrochloride is an effective and selective antagonist of protease activated receptor 1 (PAR1) with an IC50 of 70 nM and a Ki of 35 nM. SCH79797 dihydrochloride has antiproliferative and pro-apoptotic effects. purity: 98%