Learn More
TARGETMOL CHEMICALS INC SCH900776 5MG
SDP
Catalog No. 502577100
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SCH900776 (MK-8776) is an agent targeting cell cycle checkpoint kinase 1 (Chk1) with potential radiosensitization and chemosensitization activities. purity: 99%
Catalog No. 50-257-7100 Supplier TARGETMOL CHEMICALS INC Supplier No. T25175MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SCH900776 (MK-8776) is an agent targeting cell cycle checkpoint kinase 1 (Chk1) with potential radiosensitization and chemosensitization activities. purity: 99%