Learn More
TARGETMOL CHEMICALS INC SCH900776 S-ISOMER 10MG
SDP
Catalog No. 502580153
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SCH900776 S-isomer (MK-8776 S-isomer) is an effective specific and orally bioavailable inhibitor of checkpoint kinase Chk1 (IC50 3 nM). It also inhibitors Chk2 (IC50 1500 nM) and cyclin-dependent kinase CDK2 (IC50 160 nM). purity: 99%
Catalog No. 50-258-0153 Supplier TARGETMOL CHEMICALS INC Supplier No. T370010MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SCH900776 S-isomer (MK-8776 S-isomer) is an effective specific and orally bioavailable inhibitor of checkpoint kinase Chk1 (IC50 3 nM). It also inhibitors Chk2 (IC50 1500 nM) and cyclin-dependent kinase CDK2 (IC50 160 nM). purity: 99%