Learn More
Medchemexpress LLC HY-15532B 5mg Medchemexpress, SCH900776 (S-isomer) CAS:891494-64-7 Purity:>98%
SDP
Catalog No. 502081732
Encompass_Preferred
Medchemexpress, HY-15532B 5mg SCH900776 (S-isomer) CAS:891494-64-7 SCH900776 S-isomer is the S-isomer of SCH900776. SCH900776 is a potent, selective and orally bioavailable inhibitor of checkpoint kinase1 (Chk1) with IC50 of 3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1732 Supplier Medchemexpress LLC Supplier No. HY15532B5MG
Medchemexpress, HY-15532B 5mg SCH900776 (S-isomer) CAS:891494-64-7 SCH900776 S-isomer is the S-isomer of SCH900776. SCH900776 is a potent, selective and orally bioavailable inhibitor of checkpoint kinase1 (Chk1) with IC50 of 3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.