Learn More
MOLTOX Molecular Toxicology Inc Schaedler agar w/ 5% sheep's blood. 20 ml/plate. 10 plates/sleeve. Store in refrigerator.
SDP
Catalog No. NC1655959
Encompass
Schaedler Agar is a base medium for the recovery of anaerobic microorgansims. The addition of 5% sheep's blood is used for the selective isolation of the gram positive cocci. Other enrichments, such as Vitamin K1, may be added for the cultivation and isolation of fastidious aerobic and anaerobic microorganisms. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. NC1655959 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S328
Schaedler Agar is a base medium for the recovery of anaerobic microorgansims. The addition of 5% sheep's blood is used for the selective isolation of the gram positive cocci. Other enrichments, such as Vitamin K1, may be added for the cultivation and isolation of fastidious aerobic and anaerobic microorganisms. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.