Learn More
MSC Schaefer Portable Circulation Fan, 36 in, 1/2 HP, 2 Speed
Catalog No. 5000509328
Encompass
This OSHA compliant circulation fan is designed to meet all safety regulations while providing maximum airflow. It is a 2-speed fan that can be mounted on a ceiling, wall, pedestal, or cart, making it suitable for commercial, industrial, or agricultural facilities.
  • Matched direct drive motors and blades for the highest achievable airflow levels.
  • Powder-coated steel guards for corrosion resistance and durability.
  • Energy-efficient design for cost savings, drawing as little as 1.6 amps.
  • Thermally protected, heavy-duty, maintenance-free, 2-speed motors for long life.
  • Two-year warranty.
  • Made in the USA.
Catalog No. 50-005-09328 Supplier MSC Supplier No. 66991894
This OSHA compliant circulation fan is designed to meet all safety regulations while providing maximum airflow. It is a 2-speed fan that can be mounted on a ceiling, wall, pedestal, or cart, making it suitable for commercial, industrial, or agricultural facilities.
  • Matched direct drive motors and blades for the highest achievable airflow levels.
  • Powder-coated steel guards for corrosion resistance and durability.
  • Energy-efficient design for cost savings, drawing as little as 1.6 amps.
  • Thermally protected, heavy-duty, maintenance-free, 2-speed motors for long life.
  • Two-year warranty.
  • Made in the USA.