Learn More
Cayman Chemical Schaftoside, Purity: ≥98%, CAS: 51938-32-0, 10 mg
SDP
Catalog No. 502937527
Encompass_Preferred
A flavonoid C-glycoside with diverse biological activities; inhibits xanthine oxidase (IC50 = 51.4 μM) and has antioxidant activity in a β-carotene-linoleic acid bleaching assay (IC50 = 46.3 μM); inhibits neutrophil influx into mouse lung by 62% in a model of airway inflammation induced by aerosol administration of LPS at 400 μg/kg
Catalog No. 50-293-7527 Supplier Cayman Chemical Supplier No. 2795010 MG
A flavonoid C-glycoside with diverse biological activities; inhibits xanthine oxidase (IC50 = 51.4 μM) and has antioxidant activity in a β-carotene-linoleic acid bleaching assay (IC50 = 46.3 μM); inhibits neutrophil influx into mouse lung by 62% in a model of airway inflammation induced by aerosol administration of LPS at 400 μg/kg