Learn More
Oxyrase Schaedler Blood Agar Oxyplate 10/Pk
SDP
Catalog No. 503001999
Encompass
Schaedler OxyPlate patented design for anaerobe primary set-ups or subbing. Creates a seal that maintains anaerobiosis. Incubate in standard incubator. Regenerates and maintains anaerobic conditions repeatedly. Does not need/use anaerobic jars/bags/chambers. This product consists of 10 individually wrapped plates
Catalog No. 503001999 Supplier Oxyrase Supplier No. OSCWBA
Schaedler OxyPlate patented design for anaerobe primary set-ups or subbing. Creates a seal that maintains anaerobiosis. Incubate in standard incubator. Regenerates and maintains anaerobic conditions repeatedly. Does not need/use anaerobic jars/bags/chambers. This product consists of 10 individually wrapped plates