Learn More
Bioworld Schenk and Hildebrandt Medium, w/ Vitamins, 50 L
Catalog No. 50254830
Encompass_Preferred
Schenk & Hildebrandt MediumContains Macronutrients, Micronutrients, and Vitamins as described by Schenk and Hildebrandt (1972)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumWorking Concentration: 4.2 g/LpH: 3.75−4.75Soluble in water
Catalog No. 50-254-830 Supplier Bioworld Supplier No. 306301354
Schenk & Hildebrandt MediumContains Macronutrients, Micronutrients, and Vitamins as described by Schenk and Hildebrandt (1972)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumWorking Concentration: 4.2 g/LpH: 3.75−4.75Soluble in water