Learn More
Bioworld Schenk and Hildebrandt Medium, 50 L
Catalog No. 50254780
Encompass_Preferred
Schenk & Hildebrandt MediumContains Macronutrients and Micronutrients as described by Schenk and Hildebrandt (1972) Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 3.2 g/LSoluble in water
Catalog No. 50-254-780 Supplier Bioworld Supplier No. 306301114
Schenk & Hildebrandt MediumContains Macronutrients and Micronutrients as described by Schenk and Hildebrandt (1972) Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 3.2 g/LSoluble in water