Learn More
Bioworld Schenk & Hildebrandt Modified Salt Mixture w/o CaCl2, 25 L
Catalog No. 50254750
Encompass_Preferred
Schenk & Hildebrandt Modified Salt MixtureContains the Macronutrients and Micronutrients as described by Schenk & Hildebrandt (1972)Does not contain Calcium ChlorideSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 3.05 g/LSoluble in water
Catalog No. 50-254-750 Supplier Bioworld Supplier No. 306300932
Schenk & Hildebrandt Modified Salt MixtureContains the Macronutrients and Micronutrients as described by Schenk & Hildebrandt (1972)Does not contain Calcium ChlorideSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 3.05 g/LSoluble in water