Learn More
Bioworld Schenk & Hildebrandt Modified Medium, w/ Sucrose, 25 L
Catalog No. 50254935
Encompass_Preferred
Schenk & Hildebrandt Modified MediumContains Micronutrients and Macronutrients as described by Schenk & Hildebrandt (1972) with the addition of SucroseCytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 13.20 g/LSoluble in water
Catalog No. 50-254-935 Supplier Bioworld Supplier No. 306301763
Schenk & Hildebrandt Modified MediumContains Micronutrients and Macronutrients as described by Schenk & Hildebrandt (1972) with the addition of SucroseCytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 13.20 g/LSoluble in water