Learn More
Bioworld Schenk and Hildebrandt Vitamin Mix, 100 mL
Catalog No. 50254781
Encompass_Preferred
Schenk & Hildebrandt Vitamin MixContains Vitamins as described by Schenk & Hildebrandt (1972)Package contains 10.1g of vitamins to prepare 100 mL of 100X stock solution Use 10 mL of solution to prepare 1 liter of medium Contents: myo Inositol: 1000.0 mg/LNicotinic acid: 5.0 mg/LPyridoxine HCl: 0.5 mg/LThiamine HCl: 5.0 mg/L
Catalog No. 50-254-781 Supplier Bioworld Supplier No. 306301121
Schenk & Hildebrandt Vitamin MixContains Vitamins as described by Schenk & Hildebrandt (1972)Package contains 10.1g of vitamins to prepare 100 mL of 100X stock solution Use 10 mL of solution to prepare 1 liter of medium Contents: myo Inositol: 1000.0 mg/LNicotinic acid: 5.0 mg/LPyridoxine HCl: 0.5 mg/LThiamine HCl: 5.0 mg/L