Learn More
Volu Sol Schiff Reagent, 32 oz / 1 L, Volu-Sol™
SDP
Supplier:  Volu Sol VSR032
Encompass_Preferred
For use in the standard Periodic Acid-Schiff reaction for demonstration of polysaccharides, neutral mucosubstances, basement membrane, and fungal cell walls. Synonyms: Schiff's Reagent
Catalog No. 50-169-4713