Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Schiff Reagent
SCI_ED

Catalog No. S25794 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25794 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25794

missing translation for 'validMainframeMsg'

Test for aldehydes

This product is an Educational Grade Chemical — no C of A is available.

Specifications

Chemical Name or Material Schiff Reagent
CAS 64365-11-3, 7440-44-0, 7647-01-0, 7681-57-4, 7732-18-5
Assay Percent Range <1% w/v
Molecular Formula C
Synonym Test for aldehydes
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Molecular Weight (g/mol) 12.01
Packaging Solution
Quantity 500 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.