Learn More
Electron Microscopy Sciences Schiff's Reagent 1 LT
SDP
Catalog No. 5028501
Encompass_Preferred
Schiff's Reagent is used in the Periodic Acid Leucofuchsin (PAS) method as a general tissue stain.
Catalog No. 50-285-01 Supplier Electron Microscopy Sciences Supplier No. 2605206
Schiff's Reagent is used in the Periodic Acid Leucofuchsin (PAS) method as a general tissue stain.