Learn More
Electron Microscopy Sciences Schiff's Reagent, 500ml
SDP
Supplier:  Electron Microscopy Sciences 2605205
Encompass_Preferred
Schiffs reagent for the periodic acid Schiff, or PAS reaction, is used for the detection of mucopolysaccharides, glycogen, muco- and glycoproteins, glyco-, phospholipids, basal membranes and collagen. With Schiff's reagent aldehydes give a bright red color reaction, thus in combination with the periodic acid the nuclei appear blue. 500 ml are sufficient for approx. 2500 - 3000 stainings. It is registered as IVD and CE certified product and can be used for clinical diagnostics.
Catalog No. 50-285-00