Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Schiff's reagent

Catalog No. M1090330500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1090330500 500 mL
M1090332500 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1090330500 Supplier MilliporeSigma™ Supplier No. 1.09033.0500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for Microscopy

  • Used for detection of mucopolysaccharide, glycogen, muco, glycoproteins, glyco, phospholipids, basal membranes collagen
  • Schiff's reagent aldehydes give bright red color reaction nuclei appear blue
TRUSTED_SUSTAINABILITY

Specifications

Packaging Plastic Bottle
Quantity 500 mL
Storage Requirements 15° to 25°C
Product Type Schiff's Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.