Learn More
Schiff's Reagent, StatLab™
SDP
Catalog No. STL337001
Change view
Click to view available options
Quantity:
1 L
1 gal./Bottle, 4 Bottles/Cs.
16 oz.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STL337001 1 gal./Bottle, 4 Bottles/Cs.
STL3370016 16 oz.
ST337001000 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STL337001 Supplier SLMP Llc Dba Statlab Medical Supplier No. 337001
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
For use in the standard Periodic Acid-Schiff reaction.
  • For demonstration of polysaccharides, neutral mucosubstances, basement membrane, and fungal cell walls
Specifications
Quantity 1 gal./Bottle, 4 Bottles/Cs.
Product Type Schiff Reagent