Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Schiff's Reagent, StatLab™
SDP

Catalog No. STL3370016
Change view
Click to view available options
Quantity:
1 L
1 gal./Bottle, 4 Bottles/Cs.
16 oz.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STL3370016 16 oz.
ST337001000 1 L
STL337001 1 gal./Bottle, 4 Bottles/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STL3370016 Supplier SLMP Llc Dba Statlab Medical Supplier No. 3370016
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.

For use in the standard Periodic Acid-Schiff reaction.

  • For demonstration of polysaccharides, neutral mucosubstances, basement membrane, and fungal cell walls

Specifications

Quantity 16 oz.
Product Type Schiff Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.