Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Schiff's Reagent, Vion Biosciences
SCI_ED

Catalog No. S02486 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02486 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals (Fuchsin Basic) which is known by the state of California to cause cancer.
Catalog No. S02486 Supplier Aldon Corporation Supplier No. VNSS0110500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals (Fuchsin Basic) which is known by the state of California to cause cancer.

Histology grade Schiff's reagent is a versatile, ready-to-use staining solution for staining mucopolysaccharides in human cells, such as liver and intestine cells.

  • Histology grade Schiff's reagent is a versatile, ready-to-use staining solution for staining mucopolysaccharides in human cells, such as liver and intestine cells.

Specifications

Boiling Point ~100°C (212°F in water)
CAS 1-1-7647
Color Yellow-Colorless
Compatible Cells Human liver and intestine cells.
Description Clear, colorless to yellow liquid with a pungent odor.
Detection Method Mucopolysaccharide staining
For Use With (Application) Histology
Label or Dye Dye
Melting Point ~0°C (32°F in water)
Molecular Formula Mixture
Packaging Amber glass bottle
Product Line Vion Histology
Quantity 500 mL
Reagent Type Stain Solution
Sample Type Human liver and intestine cells.
Solubility Soluble in water.
Solution Type Aqueous
Storage Requirements Store in a cool, well-ventilated area away from incompatible substances. Keep away from ignition sources.
Synonym Schiff Aldehyde Solution
Target Species Human
Target Specificity Mucopolysaccharides
Product Type Schiff's Reagent
Assay Aldehyde Detection
Show More Show Less
ShelfLife 3 Years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.