Learn More
Electron Microscopy Sciences Schiff's Reagent, 500ml
SDP
Catalog No. 5030127
Encompass_Preferred
Schiffs reagent for the periodic acid Schiff, or PAS reaction, is used for the detection of mucopolysaccharides, glycogen, muco- and glycoproteins, glyco-, phospholipids, basal membranes and collagen. With Schiff's reagent aldehydes give a bright red color reaction, thus in combination with the periodic acid the nuclei appear blue. 500 ml are sufficient for approx. 2500 - 3000 stainings. It is registered as IVD and CE certified product and can be used for clinical diagnostics.
Catalog No. 50-301-27 Supplier Electron Microscopy Sciences Supplier No. 2685301
Schiffs reagent for the periodic acid Schiff, or PAS reaction, is used for the detection of mucopolysaccharides, glycogen, muco- and glycoproteins, glyco-, phospholipids, basal membranes and collagen. With Schiff's reagent aldehydes give a bright red color reaction, thus in combination with the periodic acid the nuclei appear blue. 500 ml are sufficient for approx. 2500 - 3000 stainings. It is registered as IVD and CE certified product and can be used for clinical diagnostics.