Learn More
Aviva Systems Biology Corp Schip1 Antibody - N-terminal region (ARP53777_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53777_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Schip1. It was validated on Western Blot. . Alternate names: Nf2ip, SCHIP-1, Schip-1a, Schip-1b, Schip-1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-883