Learn More
Medchemexpress LLC Schisandrin | 7432-28-2 | 432.51 | 100 MG
SDP
Catalog No. 5000415425
Encompass_Preferred
Schisandrin (Schizandrin), a dibenzocyclooctadiene lignan, is isolated from the fruit of Schisandra chinensis Baill. It exhibits various biological activities, including antioxidant, hepatoprotective, anti-cancer, and anti-inflammatory effects. This compound has also been shown to reverse memory impairment in animal models.
  • Exhibits antioxidant activity.
  • Provides hepatoprotective benefits.
  • Demonstrates anti-cancer properties.
  • Acts as an anti-inflammatory agent.
  • Reverses memory impairment in animal studies.
Catalog No. 50-004-15425 Supplier Medchemexpress LLC Supplier No. HYN0691100MG
Schisandrin (Schizandrin), a dibenzocyclooctadiene lignan, is isolated from the fruit of Schisandra chinensis Baill. It exhibits various biological activities, including antioxidant, hepatoprotective, anti-cancer, and anti-inflammatory effects. This compound has also been shown to reverse memory impairment in animal models.
  • Exhibits antioxidant activity.
  • Provides hepatoprotective benefits.
  • Demonstrates anti-cancer properties.
  • Acts as an anti-inflammatory agent.
  • Reverses memory impairment in animal studies.