Learn More
Medchemexpress LLC Schisandrin | 7432-28-2 | MFCD00905761 | 432.51 | 5 MG
SDP
Catalog No. 5000416413
Encompass_Preferred
Schisandrin (Schizandrin), a dibenzocyclooctadiene lignan, is isolated from the fruit of Schisandra chinensis Baill. Schisandrin exhibits antioxidant, hepatoprotective, anti-cancer, and anti-inflammatory activities. Schisandrin can also reverse memory impairment in rats.
  • Isolated from the fruit of Schisandra chinensis Baill.
  • Exhibits antioxidant activity.
  • Shows hepatoprotective effects.
  • Possesses anti-cancer properties.
  • Demonstrates anti-inflammatory activities.
  • Can reverse memory impairment in rats.
Catalog No. 50-004-16413 Supplier Medchemexpress LLC Supplier No. HYN06915MG
Schisandrin (Schizandrin), a dibenzocyclooctadiene lignan, is isolated from the fruit of Schisandra chinensis Baill. Schisandrin exhibits antioxidant, hepatoprotective, anti-cancer, and anti-inflammatory activities. Schisandrin can also reverse memory impairment in rats.
  • Isolated from the fruit of Schisandra chinensis Baill.
  • Exhibits antioxidant activity.
  • Shows hepatoprotective effects.
  • Possesses anti-cancer properties.
  • Demonstrates anti-inflammatory activities.
  • Can reverse memory impairment in rats.