Learn More
Medchemexpress LLC HY-N0693 10mg Medchemexpress, Schisandrin A CAS:61281-38-7 Purity:>98%
SDP
Catalog No. 501933680
Encompass_Preferred
Medchemexpress, HY-N0693 10mg Schisandrin A CAS:61281-38-7 Schisandrin A inhibits CYP3A activity with an IC 50 of 6.60 μM and K i of 5.83 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3680 Supplier Medchemexpress LLC Supplier No. HYN069310MG
Medchemexpress, HY-N0693 10mg Schisandrin A CAS:61281-38-7 Schisandrin A inhibits CYP3A activity with an IC 50 of 6.60 μM and K i of 5.83 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.