Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (+)-Schisandrin B | 82467-52-5 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN22675MG

Encompass_Preferred

(+)-Schisandrin B is an enantiomer of Schisandrin B, an active dibenzocyclooctadiene derivative isolated from the fruit of Schisandra chinensis. It exhibits an antioxidant effect on rodent liver and heart.

  • Purity: 99.56%
  • Molecular weight: 400.46
  • Formula: C23H28O6
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Phenylpropanoids, Lignans
  • Initial source: Plants, Schisandraceae, Schisandra chinensis (Turcz.) Baill.
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)
  • Solubility in DMSO: 25 mg/mL (62.43 mM)
  • Intended for research use only

Catalog No. 50-004-18717


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.