Learn More
Medchemexpress LLC Schisandrin C | 61301-33-5 | 384.42 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0690100MG
Encompass_Preferred
Schisandrin C (also known as Schizandrin-C or Wuweizisu-C) is a phytochemical lignan isolated from Schizandra chinensis. It exhibits diverse biological activities, including anticancer, anti-inflammatory, and antioxidant effects. This compound is suitable for research related to cancer, Alzheimer's disease, and liver diseases. Schisandrin C has been shown to induce cell apoptosis.
  • Isolated from Schizandra chinensis.
  • Possesses anticancer properties.
  • Offers anti-inflammatory effects.
  • Acts as an antioxidant.
  • Induces cell apoptosis.
  • Applicable for Alzheimer's disease research.
  • Used in liver diseases research.
Catalog No. 50-004-16964