Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Schisandrin C | 61301-33-5 | MFCD00078864 | 384.42 | 50 MG
SDP

Catalog No. 5000416150
Encompass_Preferred

Schisandrin C, also known as Schizandrin-C or Wuweizisu-C, is a bioactive lignan derived from *Schisandra chinensis*. It is noted for its diverse biological activities, including anticancer, anti-inflammatory, and antioxidant effects. This phytochemical induces cell apoptosis and is investigated for its potential in treating cancer, Alzheimer's disease, and liver diseases. It is a phytochemical lignan isolated from *Schizandra chinensis* and can be utilized in research pertaining to cancer, Alzheimer's disease, and liver diseases.

  • Source: Phytochemical lignan isolated from *Schizandra chinensis*.
  • Biological activities: Anticancer, anti-inflammatory, and antioxidant effects.
  • Research applications: Used in research for cancer, Alzheimer's disease, and liver diseases.
  • Mechanism of action: Induces cell apoptosis.
  • Purity: 99.95%.
  • Appearance: Solid, white to off-white.

Catalog No. 50-004-16150 Supplier Medchemexpress LLC Supplier No. HYN069050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Schisandrin C, also known as Schizandrin-C or Wuweizisu-C, is a bioactive lignan derived from *Schisandra chinensis*. It is noted for its diverse biological activities, including anticancer, anti-inflammatory, and antioxidant effects. This phytochemical induces cell apoptosis and is investigated for its potential in treating cancer, Alzheimer's disease, and liver diseases. It is a phytochemical lignan isolated from *Schizandra chinensis* and can be utilized in research pertaining to cancer, Alzheimer's disease, and liver diseases.

  • Source: Phytochemical lignan isolated from *Schizandra chinensis*.
  • Biological activities: Anticancer, anti-inflammatory, and antioxidant effects.
  • Research applications: Used in research for cancer, Alzheimer's disease, and liver diseases.
  • Mechanism of action: Induces cell apoptosis.
  • Purity: 99.95%.
  • Appearance: Solid, white to off-white.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.