Learn More
Medchemexpress LLC HY-N0694 10mg Medchemexpress, Schisantherin A CAS:58546-56-8 Purity:>98%
SDP
Catalog No. 501933681
Encompass_Preferred
Medchemexpress, HY-N0694 10mg Schisantherin A CAS:58546-56-8 Schisantherin A is a dibenzocyclooctadiene lignan isolated from the fruit of Schisandra sphenanthera. Schisantherin A inhibits p65-NF-κB translocation into the nucleus by IκBα degradation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3681 Supplier Medchemexpress LLC Supplier No. HYN069410MG
Medchemexpress, HY-N0694 10mg Schisantherin A CAS:58546-56-8 Schisantherin A is a dibenzocyclooctadiene lignan isolated from the fruit of Schisandra sphenanthera. Schisantherin A inhibits p65-NF-κB translocation into the nucleus by IκBα degradation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.