Learn More
Medchemexpress LLC Schisantherin D | 64917-82-4 | 1 MG
SDP
Catalog No. 5000419511
Encompass_Preferred
Schisantherin D is a dibenzocyclooctadiene lignan isolated from the fruit of Schisandra sphenanthera. It exhibits anti-HIV replication activities with an EC50 of 0.5 μg/mL. Additionally, Schisantherin D inhibits endothelin receptor B (ETBR) and possesses hepatoprotective effects. It is intended for research use only.
  • Exhibits anti-HIV replication activity
  • Inhibits endothelin receptor B (ETBR)
  • Possesses hepatoprotective effects
  • Isolated from Schisandra sphenanthera
  • High purity of 99.66%
Catalog No. 50-004-19511 Supplier Medchemexpress LLC Supplier No. HYN75431MG
Schisantherin D is a dibenzocyclooctadiene lignan isolated from the fruit of Schisandra sphenanthera. It exhibits anti-HIV replication activities with an EC50 of 0.5 μg/mL. Additionally, Schisantherin D inhibits endothelin receptor B (ETBR) and possesses hepatoprotective effects. It is intended for research use only.
  • Exhibits anti-HIV replication activity
  • Inhibits endothelin receptor B (ETBR)
  • Possesses hepatoprotective effects
  • Isolated from Schisandra sphenanthera
  • High purity of 99.66%