Learn More
Medchemexpress LLC Schisantherin D | 64917-82-4 | 5 MG
SDP
Catalog No. 5000419939
Encompass_Preferred
Schisantherin D is a dibenzocyclooctadiene lignan isolated from the fruit of *Schisandra sphenanthera*. It demonstrates anti-HIV replication activities with an EC50 of 0.5 μg/mL. Additionally, Schisantherin D inhibits endothelin receptor B (ETBR) and exhibits hepatoprotective effects.
  • Dibenzocyclooctadiene lignan
  • Isolated from the fruit of *Schisandra sphenanthera*
  • Demonstrates anti-HIV replication activities
  • Inhibits endothelin receptor B (ETBR)
  • Exhibits hepatoprotective effects
Catalog No. 50-004-19939 Supplier Medchemexpress LLC Supplier No. HYN75435MG
Schisantherin D is a dibenzocyclooctadiene lignan isolated from the fruit of *Schisandra sphenanthera*. It demonstrates anti-HIV replication activities with an EC50 of 0.5 μg/mL. Additionally, Schisantherin D inhibits endothelin receptor B (ETBR) and exhibits hepatoprotective effects.
  • Dibenzocyclooctadiene lignan
  • Isolated from the fruit of *Schisandra sphenanthera*
  • Demonstrates anti-HIV replication activities
  • Inhibits endothelin receptor B (ETBR)
  • Exhibits hepatoprotective effects