Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Schisantherin E | 64917-83-5 | 538.59 | 1 MG
SDP

Supplier:  Medchemexpress LLC HYN08601MG

Encompass_Preferred

Schisantherin E is a natural compound isolated from the active fraction of the fruits of Schisandra sphenanthera Rehd. et Wils. It is for research use only.

  • Purity: 99.89%
  • Appearance: Solid
  • Initial source: Plants, Schisandraceae, Schisandra chinensis (Turcz.) Baill.
  • Shipping at room temperature in continental US.
  • Store at 4°C, protected from light.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light.

Catalog No. 50-004-15985


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.