Learn More
Cayman Chemical SchIsnhenol 10mg
SDP
Catalog No. 502988915
Encompass_Preferred
A lignan with diverse biological activities; completely inhibits iron and cysteine-induced MDA formation in rat liver microsomes at 1 mM; reduces HIV-1 viral titers in H9 T cells (EC50 = 5.7 µM); protective against tobacco mosaic virus (TMV) infection in N. glutinosa when applied to leaves from 0.15-0.5 mM; prevents scopolamine-induced increases in hippocampal MDA and AChE levels and decreases hippocampal SOD and GPX activities, as well as increases time spent in the target quadrant in the Morris water maze in mice at 10, 30, or 100 mg/kg
Catalog No. 50-298-8915 Supplier Cayman Chemical Supplier No. 3010910 MG
A lignan with diverse biological activities; completely inhibits iron and cysteine-induced MDA formation in rat liver microsomes at 1 mM; reduces HIV-1 viral titers in H9 T cells (EC50 = 5.7 µM); protective against tobacco mosaic virus (TMV) infection in N. glutinosa when applied to leaves from 0.15-0.5 mM; prevents scopolamine-induced increases in hippocampal MDA and AChE levels and decreases hippocampal SOD and GPX activities, as well as increases time spent in the target quadrant in the Morris water maze in mice at 10, 30, or 100 mg/kg