Learn More
Sino Biological Schistosoma japonicum GST PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50586MNAH1
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Glutathione S-transferase (GST). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-4093