Learn More
Sino Biological Schistosoma japonicum GST PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11213RP04100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Glutathione S-transferase (GST). GST specific IgG was purified by GST affinity chromatography.
Catalog No. 50-237-1744