Learn More
Sino Biological Recombinant Schistosoma japonicum GST Protein 1
Supplier:  Sino Biological 11213HNAB1
Encompass_Preferred
A DNA sequence encoding the Glutathione S-transferase (P08515) (Met 1-Lys 218), with additional 11 amino acids at the C-terminus, was expressed and purified.
Catalog No. 50-224-6973