Learn More
Medchemexpress LLC Schizandrin-A | 61281-38-7 | 416.51 | 1 ML
SDP
Catalog No. 5000416972
Encompass_Preferred
Schisandrin A inhibits CYP3A activity with an IC50 of 6.60 μM and Ki of 5.83 μM. It is intended for research use only.
  • Inhibits CYP3A activity with an IC50 of 6.60 μM and Ki of 5.83 μM
  • Demonstrates antiproliferative activity against various human cell lines
  • Inhibits VP-16, VCR, and DNR transport in HL60/ADR cells
  • Significantly inhibits CYP3A activity in rat hepatic microsomes in vivo
Catalog No. 50-004-16972 Supplier Medchemexpress LLC Supplier No. HYN069310MM/1ML
Schisandrin A inhibits CYP3A activity with an IC50 of 6.60 μM and Ki of 5.83 μM. It is intended for research use only.
  • Inhibits CYP3A activity with an IC50 of 6.60 μM and Ki of 5.83 μM
  • Demonstrates antiproliferative activity against various human cell lines
  • Inhibits VP-16, VCR, and DNR transport in HL60/ADR cells
  • Significantly inhibits CYP3A activity in rat hepatic microsomes in vivo