Learn More
Ace Glass, Inc. Tube, reaction/storage, 25mL, 14/20 standard taper neck, 2mmglass stopcock side arm, No-Air
SDP
Catalog No. NC0169476
Encompass_Preferred
25mL Schlenk-style reaction tube for vacuum and No-Air applications. Features a 14/20 standard taper neck, and a 2mm glass stopcock side arm.
Catalog No. NC0169476 Supplier Ace Glass, Inc. Supplier No. 775208
25mL Schlenk-style reaction tube for vacuum and No-Air applications. Features a 14/20 standard taper neck, and a 2mm glass stopcock side arm.