Learn More
ILEX LIFE SCIENCES LLC SCHNEIDERS P04-91500 500 ML
SDP
Catalog No. 502733757
Encompass_Preferred
PAN-Biotech Schneider’s Drosophila Medium, w: L-Glutamine, w: 0.4 g/L NaHCO3 (500 ml). BICARBONATE: With 0.4 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With L-glutamine. PHENOL RED: Without Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: Without sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Originally developed for the culture of Drosophila cells, this medium is also suitable for the culture of other dipteran cell lines.
Catalog No. 50-273-3757 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0491500
PAN-Biotech Schneider’s Drosophila Medium, w: L-Glutamine, w: 0.4 g/L NaHCO3 (500 ml). BICARBONATE: With 0.4 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With L-glutamine. PHENOL RED: Without Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: Without sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Originally developed for the culture of Drosophila cells, this medium is also suitable for the culture of other dipteran cell lines.